2-(3-Methoxyphenyl)pyridine-3-carboxaldehyde 95+%
Lieferant: Apollo Scientific
OR346436-1G
APOSOR346436-1GEA
0
EUR
APOSOR346436-1G
2-(3-Methoxyphenyl)pyridine-3-carboxaldehyde 95+%
2-(3-Methoxyphenyl)pyridine-3-carboxaldehyde
Formel:
C₁₃H₁₁NO₂ Molecular Weight: 213,232 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD19981446 CAS-Nummer: 958219-62-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...