Benzofuran-3-carboxaldehyde 95+%

Lieferant: Apollo Scientific

Synonyme: Benzofuran-3-carbaldehyde

OR346443-1G OR346443-5G
APOSOR346443-1GEA 56.7 EUR
APOSOR346443-1G APOSOR346443-5G
Benzofuran-3-carboxaldehyde 95+%
Benzofuran-3-carboxaldehyde
Formel: C₉H₆O₂
Molecular Weight: 146.143 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03412541
CAS-Nummer: 4687-25-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR