4-(3,5-Dimethylisoxazol-4-yl)phenylamine 95+%

Lieferant: Apollo Scientific

OR346484-1G OR346484-500MG
APOSOR346484-1GEA 497 EUR
APOSOR346484-1G APOSOR346484-500MG
4-(3,5-Dimethylisoxazol-4-yl)phenylamine 95+%
4-(3,5-Dimethylisoxazol-4-yl)phenylamine
Formel: C₁₁H₁₂N₂O
Molecular Weight: 188,226 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16302749
CAS-Nummer: 2155-99-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR