2-Bromo-4-methylbenzenesulphonyl chloride 95+%
Lieferant: Apollo Scientific
OR346499-1G
OR346499-5G
APOSOR346499-1GEA
318
EUR
APOSOR346499-1G
APOSOR346499-5G
2-Bromo-4-methylbenzenesulphonyl chloride 95+%
2-Bromo-4-methylbenzenesulphonyl chloride
Formel:
C₇H₆BrClO₂S Molecular Weight: 269,543 g/mol Schmelzpunkt: 72 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08445068 CAS-Nummer: 89794-06-9 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...