1-(4-Methoxybenzyl)pyrrolidin-2-one 95+%
Lieferant: Apollo Scientific
OR346540-500MG
APOSOR346540-500MGEA
420
EUR
APOSOR346540-500MG
1-(4-Methoxybenzyl)pyrrolidin-2-one 95+%
1-(4-Methoxybenzyl)pyrrolidin-2-one
Formel:
C₁₂H₁₅NO₂ Molecular Weight: 205,253 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01960843 CAS-Nummer: 60737-65-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...