5-(2-Chloropyridin-3-yl)furan-2-carboxaldehyde 95+%

Lieferant: Apollo Scientific

OR346645-100MG OR346645-1G
APOSOR346645-100MGEA 243 EUR
APOSOR346645-100MG APOSOR346645-1G
5-(2-Chloropyridin-3-yl)furan-2-carboxaldehyde 95+%
5-(2-Chloropyridin-3-yl)furan-2-carboxaldehyde
Formel: C₁₀H₆ClNO₂
Molecular Weight: 207,613 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08898701
CAS-Nummer: 403657-26-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR