4-Bromobenzo[d]thiazol-2-amine

Lieferant: Apollo Scientific

Synonyme: 2-Amino-4-bromobenzo[d]thiazole

OR350027-5G OR350027-10G
APOSOR350027-5GEA 75.4 EUR
APOSOR350027-5G APOSOR350027-10G
4-Bromobenzo[d]thiazol-2-amine
4-Bromobenzo[d]thiazol-2-amine
Formel: C₇H₅BrN₂S
Molecular Weight: 229,097 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04971840
CAS-Nummer: 20358-02-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR