7-Nitroisoquinoline, tech

Lieferant: Apollo Scientific
Achtung

OR350298-1G OR350298-250MG
APOSOR350298-1GEA 166 EUR
APOSOR350298-1G APOSOR350298-250MG
7-Nitroisoquinoline, tech
7-Nitroisoquinoline
Formel: C₉H₆N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09264588
CAS-Nummer: 13058-73-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR