2-Bromo-5-ethoxypyridine

Lieferant: Apollo Scientific
Achtung

OR3575-1G OR3575-5G
APOSOR3575-1GEA 26.1 EUR
APOSOR3575-1G APOSOR3575-5G
2-Bromo-5-ethoxypyridine
2-Bromo-5-ethoxypyridine
Formel: C₇H₈BrNO
Molecular Weight: 202,049 g/mol
Schmelzpunkt: 47…48 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09260481
CAS-Nummer: 42834-01-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR