4-(Ethoxycarbonyl)cyclohexene-1-boronic acid, pinacol ester 96%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Ethyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)cyclohex-3-enecarboxylate
OR360085-1G
APOSOR360085-1GEA
58
EUR
APOSOR360085-1G
4-(Ethoxycarbonyl)cyclohexene-1-boronic acid, pinacol ester 96%
4-(Ethoxycarbonyl)cyclohexene-1-boronic acid, pinacol ester
Formel:
C₁₅H₂₅BO₄ Molecular Weight: 280,168 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11520546 CAS-Nummer: 1049004-32-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...