2-Cyanopyridine-5-boronic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: (6-Cyanopyridin-3-yl)boronic acid

OR360121-1G OR360121-5G
APOSOR360121-1GEA 72.5 EUR
APOSOR360121-1G APOSOR360121-5G
2-Cyanopyridine-5-boronic acid 97%
2-Cyanopyridine-5-boronic acid
Formel: C₆H₅BN₂O₂
Molecular Weight: 147.927 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10696633
CAS-Nummer: 1011722-07-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR