2-(Aminomethyl)phenyl boronic acid 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: (2-(Aminomethyl)phenyl)boronic acid

OR360234-1G OR360234-5G
APOSOR360234-1GEA 139 EUR
APOSOR360234-1G APOSOR360234-5G
2-(Aminomethyl)phenyl boronic acid 95%
2-(Aminomethyl)phenyl boronic acid
Formel: C₇H₁₀BNO₂
Molecular Weight: 150,971 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD06212997
CAS-Nummer: 248274-03-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR