6-Methoxypyridine-2-boronic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(6-Methoxypyridin-2-yl)boronic acid
OR360243-250MG
OR360243-1G
APOSOR360243-250MGEA
109
EUR
APOSOR360243-250MG
APOSOR360243-1G
6-Methoxypyridine-2-boronic acid
6-Methoxypyridine-2-boronic acid
Formel:
C₆H₈BNO₃ Molecular Weight: 152.944 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD03093332 CAS-Nummer: 372963-51-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...