(5-Cyano-2-methylphenyl)boronic acid 97%

Lieferant: Apollo Scientific

Synonyme: 5-Cyano-2-methylphenylboronic acid

OR360278-1G OR360278-5G
APOSOR360278-1GEA 56.7 EUR
APOSOR360278-1G APOSOR360278-5G
(5-Cyano-2-methylphenyl)boronic acid 97%
(5-Cyano-2-methylphenyl)boronic acid
Formel: C₈H₈BNO₂
Molecular Weight: 160.966 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17015778
CAS-Nummer: 867333-43-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR