2-Chloro-6-hydroxybenzeneboronic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: (2-Chloro-6-hydroxyphenyl)boronic acid

OR360309-100MG OR360309-1G OR360309-250MG
APOSOR360309-100MGEA 21.9 EUR
APOSOR360309-100MG APOSOR360309-1G APOSOR360309-250MG
2-Chloro-6-hydroxybenzeneboronic acid
2-Chloro-6-hydroxybenzeneboronic acid
Formel: C₆H₆BClO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD18393225
CAS-Nummer: 958646-70-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR