(3-Methylbutyl)boronic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: Isopentylboronic acid

OR360310-25G OR360310-5G
APOSOR360310-25GEA 313 EUR
APOSOR360310-25G APOSOR360310-5G
(3-Methylbutyl)boronic acid
(3-Methylbutyl)boronic acid
Formel: C₅H₁₃BO₂
Molecular Weight: 115,967 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07784365
CAS-Nummer: 98139-72-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR