[6-[(tert-Butoxycarbonyl)amino]pyridin-3-yl]boronic acid 97%

Lieferant: Apollo Scientific

Synonyme: 2-(tert-Butoxycarbonylamino)-pyridine-5-boronic acid, (6-((tert-Butoxycarbonyl)amino)pyridin-3-yl)boronic acid

OR360493-1G OR360493-5G
APOSOR360493-1GEA 114 EUR
APOSOR360493-1G APOSOR360493-5G
[6-[(tert-Butoxycarbonyl)amino]pyridin-3-yl]boronic acid 97%
[6-[(tert-Butoxycarbonyl)amino]pyridin-3-yl]boronic acid
Formel: C₁₀H₁₅BN₂O₄
Molecular Weight: 238.048 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11975411
CAS-Nummer: 883231-20-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR