2,1,3-Benzoxadiazole-5-carbonyl chloride 95+%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 5-(Chlorocarbonyl)-2,1,3-benzoxadiazole, Benzofurazan-5-carbonyl chloride, 5-(Chloroformyl)-2,1,3-benzoxadiazole

OR3608-1G OR3608-250MG OR3608-5G
APOSOR3608-1GEA 125 EUR
APOSOR3608-1G APOSOR3608-250MG APOSOR3608-5G
2,1,3-Benzoxadiazole-5-carbonyl chloride 95+%
2,1,3-Benzoxadiazole-5-carbonyl chloride
Formel: C₇H₃ClN₂O₂
Molecular Weight: 182,564 g/mol
Schmelzpunkt: 53…58 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00276978
CAS-Nummer: 126147-86-2
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR