2-Chloro-4-formylphenylboronic acid 98%

Lieferant: Apollo Scientific

OR360934-1G OR360934-250MG
APOSOR360934-1GEA 136 EUR
APOSOR360934-1G APOSOR360934-250MG
2-Chloro-4-formylphenylboronic acid 98%
2-Chloro-4-formylphenylboronic acid
Formel: C₇H₆BClO₃
Molecular Weight: 184,385 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18311828
CAS-Nummer: 1063712-34-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR