4-Chloro-2-methylphenylboronic acid, pinacol ester 95+%
Lieferant: Apollo Scientific
Synonyme:
2-(4-Chloro-2-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-Chloro-2-methylphenylboronic acid pinacol ester
OR360972-5G
APOSOR360972-5GEA
55.1
EUR
APOSOR360972-5G
4-Chloro-2-methylphenylboronic acid, pinacol ester 95+%
4-Chloro-2-methylphenylboronic acid, pinacol ester
Formel:
C₁₃H₁₈BClO₂ Molecular Weight: 252,545 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18729895 CAS-Nummer: 1030832-75-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...