3-Cyano-2-methylphenylboronic acid 97%

Lieferant: Apollo Scientific
Achtung

OR361032-1G OR361032-5G
APOSOR361032-1GEA 219 EUR
APOSOR361032-1G APOSOR361032-5G
3-Cyano-2-methylphenylboronic acid 97%
3-Cyano-2-methylphenylboronic acid
Formel: C₈H₈BNO₂
Molecular Weight: 160,966 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18394417
CAS-Nummer: 313546-19-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR