4-Cyano-3-methylphenylboronic acid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (4-Cyano-3-methylphenyl)boronic acid

OR361033-250MG OR361033-100MG
APOSOR361033-250MGEA 168 EUR
APOSOR361033-250MG APOSOR361033-100MG
4-Cyano-3-methylphenylboronic acid 98%
4-Cyano-3-methylphenylboronic acid
Formel: C₈H₈BNO₂
Molecular Weight: 160,966 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD18394419
CAS-Nummer: 856255-58-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR