2,3-Dihydro-5-furylboronic acid, pinacol ester 98%

Lieferant: Apollo Scientific
Achtung

OR361130-1G OR361130-5G
APOSOR361130-1GEA 453 EUR
APOSOR361130-1G APOSOR361130-5G
2,3-Dihydro-5-furylboronic acid, pinacol ester 98%
2,3-Dihydro-5-furylboronic acid, pinacol ester
Formel: C₁₀H₁₇BO₃
Molecular Weight: 196,051 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11656065
CAS-Nummer: 1046812-02-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR