3,4-Dihydro-2H-pyran-6-boronic acid, pinacol ester 95%

Lieferant: Apollo Scientific

OR361133-1G OR361133-25G OR361133-5G
APOSOR361133-1GEA 69.6 EUR
APOSOR361133-1G APOSOR361133-25G APOSOR361133-5G
3,4-Dihydro-2H-pyran-6-boronic acid, pinacol ester 95%
3,4-Dihydro-2H-pyran-6-boronic acid, pinacol ester
Formel: C₁₁H₁₉BO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD07368233
CAS-Nummer: 1025707-93-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR