5-Formylfuran-2-boronic acid, pinacol ester 95%

Lieferant: Apollo Scientific

Synonyme: 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)furan-2-carbaldehyde

OR361230-5G
APOSOR361230-5GEA 26.1 EUR
APOSOR361230-5G
5-Formylfuran-2-boronic acid, pinacol ester 95%
5-Formylfuran-2-boronic acid, pinacol ester
Formel: C₁₁H₁₅BO₄
Molecular Weight: 222,045 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16295105
CAS-Nummer: 273731-82-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR