Iodomethylboronic acid, pinacol ester 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(Iodomethyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

OR361281-5G OR361281-1G OR361281-10G
APOSOR361281-5GEA 30.6 EUR
APOSOR361281-5G APOSOR361281-1G APOSOR361281-10G
Iodomethylboronic acid, pinacol ester 95%
Iodomethylboronic acid, pinacol ester
Formel: C₇H₁₄BIO₂
Molecular Weight: 267,9 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD12405515
CAS-Nummer: 70557-99-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR