6-Bromoquinoline-3-carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Bromo-3-carboxy-1-azanaphthalene, 6-Bromo-3-carboxyquinoline

OR3615-1G
APOSOR3615-1GEA 106 EUR
APOSOR3615-1G
6-Bromoquinoline-3-carboxylic acid
6-Bromoquinoline-3-carboxylic acid
Formel: C₁₀H₆BrNO₂
Molecular Weight: 252,064 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08437553
CAS-Nummer: 798545-30-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR