1-Phenylpyrazole-4-boronic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: (1-Phenyl-1H-pyrazol-4-yl)boronic acid

OR361529-250MG OR361529-1G
APOSOR361529-250MGEA 172 EUR
APOSOR361529-250MG APOSOR361529-1G
1-Phenylpyrazole-4-boronic acid
1-Phenylpyrazole-4-boronic acid
Formel: C₉H₉BN₂O₂
Molecular Weight: 187,991 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11044914
CAS-Nummer: 1201643-70-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR