2-Piperidinopyridine-4-boronic acid 95%

Lieferant: Apollo Scientific
Achtung

OR361548-1G OR361548-250MG
APOSOR361548-1GEA 812 EUR
APOSOR361548-1G APOSOR361548-250MG
2-Piperidinopyridine-4-boronic acid 95%
2-Piperidinopyridine-4-boronic acid
Formel: C₁₀H₁₅BN₂O₂
Molecular Weight: 206.049 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17015779
CAS-Nummer: 1256358-82-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR