1-(tert-Butoxycarbonyl)pyrrole-3-boronic acid 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(1-(tert-Butoxycarbonyl)-1H-pyrrol-3-yl)boronic acid
OR361609-1G
OR361609-250MG
APOSOR361609-1GEA
1040
EUR
APOSOR361609-1G
APOSOR361609-250MG
1-(tert-Butoxycarbonyl)pyrrole-3-boronic acid 98%
1-(tert-Butoxycarbonyl)pyrrole-3-boronic acid
Formel:
C₉H₁₄BNO₄ Molecular Weight: 211,023 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD08234379 CAS-Nummer: 832697-40-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...