1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)cyclohex-3-enyl)ethanone

Lieferant: Apollo Scientific
Achtung

OR361640-1G OR361640-250MG
APOSOR361640-1GEA 1080 EUR
APOSOR361640-1G APOSOR361640-250MG
1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)cyclohex-3-enyl)ethanone
1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)cyclohex-3-enyl)ethanone
Formel: C₁₄H₂₃BO₃
Molecular Weight: 250,142 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18383335
CAS-Nummer: 151075-23-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR