4-Trimethylsilylphenylboronic acid, pinacol ester 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Trimethyl(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)silane
OR361680-5G
APOSOR361680-5GEA
197
EUR
APOSOR361680-5G
4-Trimethylsilylphenylboronic acid, pinacol ester 98%
4-Trimethylsilylphenylboronic acid, pinacol ester
Formel:
C₁₅H₂₅BO₂Si Molecular Weight: 276,254 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09037796 CAS-Nummer: 1186026-67-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...