4-Amino-2-(tert-butyl)phenol ≥98%

Lieferant: Apollo Scientific

OR361725-100MG OR361725-250MG
APOSOR361725-100MGEA 307 EUR
APOSOR361725-100MG APOSOR361725-250MG
4-Amino-2-(tert-butyl)phenol ≥98%
4-Amino-2-(tert-butyl)phenol
Formel: C₁₀H₁₅NO
Molecular Weight: 165,232 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16996828
CAS-Nummer: 4151-62-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR