(2S,3S)-2-Phenylpyrrolidine-3-carboxylic acid ≥98%

Lieferant: Apollo Scientific

OR361745-100MG OR361745-250MG
APOSOR361745-100MGEA 1080 EUR
APOSOR361745-100MG APOSOR361745-250MG
(2S,3S)-2-Phenylpyrrolidine-3-carboxylic acid ≥98%
(2S,3S)-2-Phenylpyrrolidine-3-carboxylic acid
Formel: C₁₁H₁₃NO₂
Molecular Weight: 191,226 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07374328
CAS-Nummer: 1241681-66-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR