6-Bromo-2-iminocoumarin-3-carbonitrile
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Bromo-3-cyano-2-iminocoumarin, 6-Bromo-2-imino-2H-chromene-3-carbonitrile
OR3623-1G
OR3623-250MG
OR3623-5G
APOSOR3623-1GEA
519
EUR
APOSOR3623-1G
APOSOR3623-250MG
APOSOR3623-5G
6-Bromo-2-iminocoumarin-3-carbonitrile
6-Bromo-2-iminocoumarin-3-carbonitrile
Formel:
C₁₀H₅BrN₂O Molecular Weight: 249.063 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00115365 CAS-Nummer: 860789-92-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...