8-Chloro-4-hydroxy-6-methylquinoline

Lieferant: Apollo Scientific

Synonyme: 8-Chloro-6-methylquinolin-4-ol, 8-Chloro-4-hydroxy-6-methyl-1-azanaphthalene

OR3639-1G OR3639-250MG
APOSOR3639-1GEA 284 EUR
APOSOR3639-1G APOSOR3639-250MG
8-Chloro-4-hydroxy-6-methylquinoline
8-Chloro-4-hydroxy-6-methylquinoline
Formel: C₁₀H₈ClNO
Molecular Weight: 193,63 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00272441
CAS-Nummer: 203626-40-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR