Fmoc-p-azido-D-Phe-OH 99%

Lieferant: Apollo Scientific

OR36432-1G OR36432-250MG
APOSOR36432-1GEA 551 EUR
APOSOR36432-1G APOSOR36432-250MG
Fmoc-p-azido-D-Phe-OH 99%
Fmoc-p-azido-D-Phe-OH
Formel: C₂₄H₂₀N₄O₄
Molecular Weight: 428,44 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11041252
CAS-Nummer: 1391586-30-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR