4-Amino-8-bromo-2-methylquinoline

Lieferant: Apollo Scientific

OR3655-1G OR3655-500MG
APOSOR3655-1GEA 317 EUR
APOSOR3655-1G APOSOR3655-500MG
4-Amino-8-bromo-2-methylquinoline
4-Amino-8-bromo-2-methylquinoline
Formel: C₁₀H₉BrN₂
Molecular Weight: 237,096 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08669717
CAS-Nummer: 288151-51-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR