4-Amino-6-methoxyquinoline 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Methoxyquinolin-4-amine, 4-Amino-6-methoxy-1-azanaphthalene

OR3656-1G OR3656-100MG
APOSOR3656-1GEA 260 EUR
APOSOR3656-1G APOSOR3656-100MG
4-Amino-6-methoxyquinoline 98%
4-Amino-6-methoxyquinoline
Formel: C₁₀H₁₀N₂O
Molecular Weight: 174,199 g/mol
Schmelzpunkt: 119…123 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00473716
CAS-Nummer: 6279-51-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR