Diethyl 2-amino-1,3-thiazole-4,5-dicarboxylate 95%

Lieferant: Apollo Scientific

OR370001-1G OR370001-250MG
APOSOR370001-1GEA 560 EUR
APOSOR370001-1G APOSOR370001-250MG
Diethyl 2-amino-1,3-thiazole-4,5-dicarboxylate 95%
Diethyl 2-amino-1,3-thiazole-4,5-dicarboxylate
Formel: C₉H₁₂N₂O₄S
Lagertemperatur: Kühlschrank
MDL: MFCD00570752
CAS-Nummer: 5445-93-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR