5-Methoxy-1,2,3,4-tetrahydronaphthalen-2-ol ≥98%

Lieferant: Apollo Scientific

OR370006-100MG OR370006-1G OR370006-250MG
APOSOR370006-100MGEA 219 EUR
APOSOR370006-100MG APOSOR370006-1G APOSOR370006-250MG
5-Methoxy-1,2,3,4-tetrahydronaphthalen-2-ol ≥98%
5-Methoxy-1,2,3,4-tetrahydronaphthalen-2-ol
Formel: C₁₁H₁₄O₂
Molecular Weight: 178,228 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD25969866
CAS-Nummer: 32920-08-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR