3-Cyano-3-methylcyclohexanone ≥98%

Lieferant: Apollo Scientific

OR370042-100MG OR370042-250MG
APOSOR370042-100MGEA 415 EUR
APOSOR370042-100MG APOSOR370042-250MG
3-Cyano-3-methylcyclohexanone ≥98%
3-Cyano-3-methylcyclohexanone
Formel: C₈H₁₁NO
Molecular Weight: 137,179 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24688472
CAS-Nummer: 33235-14-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR