4-Methyl-2,3-dihydrocyclopenta[b]indol-1(4H)-one ≥98%

Lieferant: Apollo Scientific

OR370045-1G OR370045-5G
APOSOR370045-1GEA 307 EUR
APOSOR370045-1G APOSOR370045-5G
4-Methyl-2,3-dihydrocyclopenta[b]indol-1(4H)-one ≥98%
4-Methyl-2,3-dihydrocyclopenta[b]indol-1(4H)-one
Formel: C₁₂H₁₁NO
Molecular Weight: 185,222 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09032316
CAS-Nummer: 50776-26-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR