1-(3,5-Dichloropyridin-4-yl)ethanol ≥98%
Lieferant: Apollo Scientific
OR370074-1G
APOSOR370074-1GEA
104
EUR
APOSOR370074-1G
1-(3,5-Dichloropyridin-4-yl)ethanol ≥98%
1-(3,5-Dichloropyridin-4-yl)ethanol
Formel:
C₇H₇Cl₂NO Molecular Weight: 192,043 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23163275 CAS-Nummer: 1254473-66-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...