2,5-Dimethyl-1,3-oxazole-4-carboxaldehyde 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,5-Dimethyl-4-formyl-1,3-oxazole, 2,5-Dimethyloxazole-4-carbaldehyde
OR3802-1G
OR3802-250MG
APOSOR3802-1GEA
429
EUR
APOSOR3802-1G
APOSOR3802-250MG
2,5-Dimethyl-1,3-oxazole-4-carboxaldehyde 97%
2,5-Dimethyl-1,3-oxazole-4-carboxaldehyde
Formel:
C₆H₇NO₂ Molecular Weight: 125,125 g/mol Siedepunkt: 38…40 °C (0,2 mm Hg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD03923827 CAS-Nummer: 92901-88-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...