3-(Hydroxymethyl)-1-methyl-1H-indazole 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: (1-Methyl-1H-indazol-3-yl)methanol

OR3815-1G OR3815-250MG
APOSOR3815-1GEA 102 EUR
APOSOR3815-1G APOSOR3815-250MG
3-(Hydroxymethyl)-1-methyl-1H-indazole 97%
3-(Hydroxymethyl)-1-methyl-1H-indazole
Formel: C₉H₁₀N₂O
Molecular Weight: 162,189 g/mol
Siedepunkt: 128 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08060470
CAS-Nummer: 1578-96-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR