1-Azabicyclo[2.2.2]octane-4-carbonitrile 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 4-Cyanonquinuclidine, Quinuclidine-4-carbonitrile, 4-Cyano-1-azabicyclo[2.2.2]octane

OR3931-1G
APOSOR3931-1GEA 78.3 EUR
APOSOR3931-1G
1-Azabicyclo[2.2.2]octane-4-carbonitrile 97%
1-Azabicyclo[2.2.2]octane-4-carbonitrile
Formel: C₈H₁₂N₂
Molecular Weight: 136,194 g/mol
Schmelzpunkt: 129…130 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00276610
CAS-Nummer: 26458-78-6
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR