2-Methoxybenzeneboronic acid, propanediol cyclic ester 97%
Lieferant: Apollo Scientific
Synonyme:
2-(2-Methoxyphenyl)-1,3,2-dioxaborinane
OR3951-5G
APOSOR3951-5GEA
84.1
EUR
APOSOR3951-5G
2-Methoxybenzeneboronic acid, propanediol cyclic ester 97%
2-Methoxybenzeneboronic acid, propanediol cyclic ester
Formel:
C₁₀H₁₃BO₃ Molecular Weight: 192,019 g/mol Schmelzpunkt: 56…60 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06659934 CAS-Nummer: 141522-26-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...