2-Bromo-6-nitrophenylacetonitrile

Lieferant: Apollo Scientific

OR399025-1G OR399025-250MG
APOSOR399025-1GEA 393 EUR
APOSOR399025-1G APOSOR399025-250MG
2-Bromo-6-nitrophenylacetonitrile
2-Bromo-6-nitrophenylacetonitrile
Formel: C₈H₅BrN₂O₂
Molecular Weight: 241,042 g/mol
Schmelzpunkt: 61…62 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD18392323
CAS-Nummer: 105018-28-8
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR