6-Bromoisatoic anhydride

Lieferant: Apollo Scientific
Gefahr

Synonyme: 5-Bromo-1H-benzo[d][1,3]oxazine-2,4-dione

OR400005-1G OR400005-250MG OR400005-5G
APOSOR400005-1GEA 29 EUR
APOSOR400005-1G APOSOR400005-250MG APOSOR400005-5G
6-Bromoisatoic anhydride
6-Bromoisatoic anhydride
Formel: C₈H₄BrNO₃
Molecular Weight: 242,026 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15526667
CAS-Nummer: 77603-45-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR